Human Recombinant IFN-A 2A (from E. coli)
Supplier: VWR International
77751-582 77751-584 77751-586 77751-588 77751-590
77751-582EA 239.88 USD
77751-582 77751-584 77751-586 77751-588 77751-590
Human Recombinant IFN-A 2A (from E. coli)
Proteins and Peptides
Interferon-alpha 2a (IFN-α 2a) is a type I interferon made by leukocytes during viral infection. The JAK-STAT pathway mediates the antiviral and anti-cell proliferation activities of IFN-α 2a. IFN-α proteins are widely used as standard treatments during antiviral and antineoplastic therapies. The IFN-α 2a variant differs from IFN-α 2b by one amino acid.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.