Human Recombinant TNFR-1 (from E. coli)
Supplier: VWR International
77751-386 77751-388 77751-390 77751-392 77751-394
77751-386EA 600.95 USD
77751-386 77751-388 77751-390 77751-392 77751-394
Human Recombinant TNFR-1 (from E. coli)
Proteins and Peptides
Tumor necrosis factor receptor type 1 (TNFR-1) is expressed in most tissues and is activated by soluble and membrane-bound tumor necrosis factor alpha (TNF-α). TNFR-1 activates NF-κB and MAPK pathways to induce inflammation, promote apoptotic cell death, inhibit tumorigenesis, and inhibit viral replication.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.