Human Recombinant FGF-Basic 147 (from E. coli)
Supplier: VWR International
77751-272 77751-274 77751-276 77751-278 77751-280
77751-272EA 434.3 USD
77751-272 77751-274 77751-276 77751-278 77751-280
Human Recombinant FGF-Basic 147 (from E. coli)
Proteins and Peptides
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Degredation of the full length FGF-basic N-terminus results in a truncated FGF-basic 147aa protein, when the protein is isolated from biological sources. The N-terminus extensions influence the localization of FGF-basic within the cell, but do not affect the biological activity of FGF-basic. Therefore, there are no detectable differences in biological activity between the full length FGF-basic 154aa and the truncated FGF-basic 147aa recombinant proteins.
  • High quality
  • Low endotoxin
  • Affordable
Caution:
For research use only.