Human Recombinant EGF (from E. coli cells)
Supplier: VWR International
77751-254 77751-256 77751-258 77751-260
77751-254EA 103.53 USD
77751-254 77751-256 77751-258 77751-260
Human Recombinant EGF (from E. coli cells)
Proteins and Peptides
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR). EGF is overexpressed in multiple tumor cell lines and promotes resistance to chemotherapy and radiation treatments.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.