Human Recombinant BAFF (from E. coli)
Supplier: VWR International
77751-214 77751-206 77751-208 77751-212
77751-214EA 95.95 USD
77751-214 77751-206 77751-208 77751-212
Human Recombinant BAFF (from E. coli)
Proteins and Peptides
B cell-activating factor (BAFF), or B lymphocyte stimulator (BLyS), is a type II member of the tumor necrosis factor (TNF) superfamily. BAFF is expressed as a transmembrane protein on T cells, macrophages, and dendritic cells. The transmembrane domain of BAFF can also be cleaved to produce a soluble protein fragment. BAFF binds to the TNF receptors known as B cell maturation antigen (BCMA), transmembrane activator and CAML interactor (TACI), and BAFF receptor (BAFFR). BAFF is important for the survival and maturation of peripheral B cells. Human BAFF shows activity on mouse splenocytes.
  • High quality
  • Low endotoxin
  • Affordable
Caution:
For research use only.