Human Recombinant IL-31 (from E. coli)
Supplier: VWR International
77751-028 77751-030 77751-032 77751-034 77751-036
77751-028EA 1199.38 USD
77751-028 77751-030 77751-032 77751-034 77751-036
Human Recombinant IL-31 (from E. coli)
Proteins and Peptides
Interleukin 31 (IL-31) is an immunoregulatory cytokine that is expressed by activated type 2 T helper (Th2) cells. IL-31 signals through a heterodimer receptor consisting of the IL-31 Receptor A (IL-31RA) and the oncostatin M receptor (OSMR), which are expressed on monocytes, epithelial cells, and keratinocytes. IL-31 promotes allergic reactions and inflammatory skin diseases.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.