Human Recombinant MIP-1 A (from E. coli)
Supplier: VWR International
77750-482 77750-492 77750-494 77750-536 77750-538
77750-482EA 600.95 USD
77750-482 77750-492 77750-494 77750-536 77750-538
Human Recombinant MIP-1 A (from E. coli)
Proteins and Peptides
Macrophage inflammatory protein-1 alpha (MIP-1 α), also known as CCL3, is a cytokine produced by macrophages.  MIP-1 α binds the chemokine receptors CCR1, CCR4 and CCR5 to induce inflammatory responses, including the recruitment of granulocytes and neutrophil superoxide production.  The MIP-1 α and MIP-1 β heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1). 
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.