Human Recombinant SCF (from E. coli)
Supplier: VWR International
Stem cell factor (SCF) is a cytokine made by fibroblasts and endothelial cells. SCF binds to the receptor c-Kit/CD117 and plays a critical role in the maintenance, survival, and differentiation of hematopoietic stem cells. While human SCF shows no activity on murine cells, murine and rat SCF are active on human cells.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.