2-Chloro-4-iodoaniline ≥98%

Supplier: Ambeed
Danger

A829317-5G
77749-619EA 11 USD
77749-619
2-Chloro-4-iodoaniline ≥98%
2-Chloro-4-iodoaniline

Caution: for Research Use Only

Formula: C₆H₅ClIN
MW: 253.47
Boiling Pt: 289
Melting Pt: 73
MDL Number: MFCD00025298
CAS Number: 42016-93-3
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR