3-Sulfopropyl methacrylate potassium salt ≥97%

Supplier: Ambeed
Warning

A560221-25G
77749-157EA 32 USD
77749-157
3-Sulfopropyl methacrylate potassium salt ≥97%
3-Sulfopropyl methacrylate potassium salt

Caution: for Research Use Only

Formula: C₇H₁₂O₅S
MDL Number: MFCD00010139
CAS Number: 31098-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR