5-Amino-2-fluorobenzoic acid 98%

Supplier: Ambeed
Warning

A199313-1G
103847-716EA 17 USD
103847-716 77748-263
5-Amino-2-fluorobenzoic acid 98%
5-Amino-2-fluorobenzoic acid

Caution: Keep in a dark place, inert atmosphere, room temperature

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 190…191 °C
Flash Pt: 166.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00077449
CAS Number: 56741-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR