1-Allylpiperazine ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with TBC

A494742-5G
77747-013EA 36 USD
77747-013
1-Allylpiperazine ≥98% stabilized
1-Allylpiperazine

Caution: for Research Use Only

Formula: C₇H₁₄N₂
MDL Number: MFCD00167970
CAS Number: 13961-36-9
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR