5(6)-Carboxy-X-rhodamine 98+% (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: 5(6)-ROX

A334877-10MG
77746-653EA 162 USD
77746-653
5(6)-Carboxy-X-rhodamine 98+% (mixture of isomers)
5(6)-Carboxy-X-rhodamine

Caution: for Research Use Only

Formula: C₃₃H₃₀N₂O₅
MDL Number: MFCD01862979
CAS Number: 198978-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR