1,7-Octadiyne 98%

Supplier: Ambeed
Danger

Synonyms: Octa-1,7-diyne

A251352-5G A251352-1G A251352-25G
77193-650EA 91 USD
77193-650 77616-046 77746-396
1,7-Octadiyne 98%
1,7-Octadiyne

Caution: for Research Use Only

Formula: C₈H₁₀
MDL Number: MFCD00008580
CAS Number: 871-84-1
UN: 3295
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR