1-Pyreneboronic acid 97%

Supplier: Ambeed
Warning

A109255-100G A109255-10G A109255-250MG A109255-25G A109255-1G A109255-5G A109255-500G
103797-476EA 9 USD
103797-476 103823-270 77233-444 77233-446 77233-448 77233-450 77310-096 77310-098 77745-814
1-Pyreneboronic acid 97%
1-Pyreneboronic acid

Caution: for Research Use Only

Formula: C₁₆H₁₁BO₂
Boiling Pt: 509.4 °C (760 mm Hg)
Melting Pt: 247…251 °C
Flash Pt: 261.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04974062
CAS Number: 164461-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR