CC-401 hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(3-(2-(Piperidin-1-yl)ethoxy)phenyl)-5-(1H-1,2,4-triazol-5-yl)-1H-indazole hydrochloride, CC-401 HCl, CC401
A101734-2MG
A101734-5MG
77233-088EA
35
USD
77233-088
77745-776
CC-401 hydrochloride 98%
CC-401 hydrochloride
Caution: for Research Use Only
Formula:
C₂₂H₂₄N₆O·HCl |
MDL Number:
MFCD21364765 CAS Number: 1438391-30-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...