Anti-NOX4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A29880-100UL
77744-720EA 585 USD
77744-720
Anti-NOX4 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR