Anti-NOV Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A29867-50UL A29867-100UL
77744-697EA 225 USD
77744-697 77744-696
Anti-NOV Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: NOV
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR