Anti-NKX3-2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A29745-100UG A29745-50UG
77744-499EA 330 USD
77744-499 77744-500
Anti-NKX3-2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:5000

Type:
Antigen: NKX3-2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR