Anti-NCOA4 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A29450-50UL A29450-100UL
77744-047EA 225 USD
77744-047 77744-046
Anti-NCOA4 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200

Type:
Antigen: NCOA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR