Anti-NCOA1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A29445-50UG A29445-100UG
77744-038EA 225 USD
77744-038 77744-037
Anti-NCOA1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: NCOA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR