Anti-MMP9 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A28707-100UL
77743-262EA 585 USD
77743-262
Anti-MMP9 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR