Anti-MAP4K3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A28107-100UL
77742-709EA 585 USD
77742-709
Anti-MAP4K3 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR