Anti-HIST1H3A (phospho-Thr6) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24955-100UL A24955-50UL
77739-897EA 350 USD
77739-897 77857-170
Anti-HIST1H3A (phospho-Thr6) Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF:1:10 to 1:100

Type:
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR