Anti-GPR101 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A23669-100UL
77738-775EA 585 USD
77738-775
Anti-GPR101 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: GPR101
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR