Anti-GCA Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A22892-100UL A22892-50UL
77738-050EA 330 USD
77738-050 77855-847
Anti-GCA Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: GCA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR