Anti-GRM6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A22491-50UG
77737-676EA 225 USD
77737-676
Anti-GRM6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:20000

Type:
Antigen: GRM6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR