Anti-ERBB2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A21309-50UG
77736-557EA 225 USD
77736-557
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:200 to 1:1000, ELISA: 1:20000

Type:
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR