Anti-ERBB2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A21306-50UG
77736-554EA 225 USD
77736-554
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:10000

Type:
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR