Anti-EFNB1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A21060-50UG
77736-328EA 225 USD
77736-328
Anti-EFNB1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:20000

Type:
Antigen: EFNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR