Anti-ASB9 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A65386-100UG A65386-50UG
77725-820EA 334.95 USD
77725-820 77725-821
Anti-ASB9 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ASB9
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR