Anti-MRAP Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A65162-100UG A65162-50UG
77725-394EA 334.95 USD
77725-394 77725-395
Anti-MRAP Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MRAP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR