Anti-HMGCLL1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A65142-100UG A65142-50UG
77725-356EA 334.95 USD
77725-356 77725-357
Anti-HMGCLL1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: HMGCLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR