Anti-STXBP6 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A65121-100UG A65121-50UG
77725-317EA 334.95 USD
77725-317 77725-318
Anti-STXBP6 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: STXBP6
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR