Anti-C12orf43 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A64824-50UG A64824-100UG
77724-754EA 228.38 USD
77724-754 77724-753
Anti-C12orf43 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: C12orf43
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR