Anti-SLC7A6 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A64735-50UG A64735-100UG
77724-581EA 228.38 USD
77724-581 77724-580
Anti-SLC7A6 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC7A6
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR