Anti-LHFPL5 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A64060-50UG A64060-100UG
77723-314EA 228.38 USD
77723-314 77723-313
Anti-LHFPL5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: LHFPL5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR