Anti-NFKBID Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A64049-50UG A64049-100UG
77723-293EA 228.38 USD
77723-293 77723-292
Anti-NFKBID Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NFKBID
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR