Anti-MARVELD3 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63651-50UG A63651-100UG
77722-537EA 228.38 USD
77722-537 77722-536
Anti-MARVELD3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: MARVELD3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR