Anti-WBP2 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63639-100UG A63639-50UG
77722-513EA 334.95 USD
77722-513 77722-514
Anti-WBP2 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: WBP2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR