Anti-ALDH8A1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63625-50UL A63625-100UL
77722-488EA 228.38 USD
77722-488 77722-487
Anti-ALDH8A1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, WB:1:1000-1:5000, IHC:1:50-1:200

Type:
Antigen: ALDH8A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR