Anti-HGD Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63623-50UL A63623-100UL
77722-484EA 228.38 USD
77722-484 77722-483
Anti-HGD Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: HGD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR