Anti-CDS1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A63619-100UG A63619-50UG
77722-475EA 334.95 USD
77722-475 77722-476
Anti-CDS1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CDS1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR