Anti-KLK6 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A63614-50UG A63614-100UG
77722-466EA 228.38 USD
77722-466 77722-465
Anti-KLK6 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: KLK6
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR