Anti-HOXB13 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63605-100UG A63605-50UG
77690-296EA 334.95 USD
77690-296 77722-448
Anti-HOXB13 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: HOXB13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR