Anti-FAM45A Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A63448-100UG A63448-50UG
77722-153EA 334.95 USD
77722-153 77722-154
Anti-FAM45A Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: FAM45A
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR