Anti-NR1H4 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A63307-100UG A63307-50UG
77721-886EA 334.95 USD
77721-886 77721-887
Anti-NR1H4 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: NR1H4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR