Anti-LSM10 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A62558-100UG A62558-50UG
77720-461EA 334.95 USD
77720-461 77720-462
Anti-LSM10 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: LSM10
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR