Anti-Ferritin-2, chloroplastic Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A62543-50UG A62543-100UG
77720-433EA 228.38 USD
77720-433 77720-432
Anti-Ferritin-2, chloroplastic Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Ferritin-2, chloroplastic
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Glycine max
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR