Anti-FAM160A2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A62045-50UG A62045-100UG
77719-550EA 228.38 USD
77719-550 77719-549
Anti-FAM160A2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: FAM160A2
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR